Learn More
R&D Systems™ Cultrex RGF Basement Membrane Extract, Type R1
Description
- Avoid freeze-thaw cycles
- Basement membranes are continuous sheets of specialized extracellular matrix that form an interface between endothelial, epithelial, muscle, or neuronal cells and their adjacent stroma.
- Basement membranes are degraded and regenerated during development and wound healing.
- They not only support cells and cell layers, but they also play an essential role in tissue organization that affects cell adhesion, migration, proliferation, and differentiation.
- Basement membranes provide major barriers to invasion by metastatic tumor cells.
- Has a higher concentration of entactin, one of the BME components that connects laminins and collagens reinforcing the hydrogel structure.
- Specifically designed to culture tissue organoids.
Specifications
Specifications
| Concentration | 8-12 mg/mL (by Lowry assay) |
| Quantity | 5 mL |
| Content And Storage | Product is stable for a minimum of 3 months from date of shipment when stored at -20° C in a manual defrost freezer. For optimal stability, store at -80° C. Avoid freeze-thaw cycles. |
| Description | For optimal stability, store at -80°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.