Learn More
R&D Systems™ Cultrex Stem Cell Qualified Laminin I
SDP
Catalog No. 340001003 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
34-000-1003 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 34-000-1003 Supplier R&D Systems™ Supplier No. 340001003
Add to Cart
Add to Cart
This item is not returnable. View return policy
R&D Systems™ Cultrex Stem Cell Qualified Laminin I provides a functionally defined and effective feeder-free surface for the attachment and maintenance of embryonic stem cells in a pluripotent state, thereby enabling its use for growth promotion or study of stem cell differentiation.
Specifications
Product Line R&D Systems
Quantity 1 mg
Content And Storage Store at -80°C in a manual defrost freezer