Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Culture Tube Caps

Catalog No. MP07DP1056 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Culture Tube Caps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MP07DP1056 Culture Tube Caps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP07DP1056 Supplier MP Biomedicals™ Supplier No. 07DP1056

missing translation for 'validMainframeMsg'

MP Biomedicals™ Culture Tube Caps to fit 10mm x 75mm culture tubes

  • Made of polypropylene
TRUSTED_SUSTAINABILITY

Specifications

Material Polypropylene
Disposable Single-use
For Use With 10 x 75mm culture tubes
Thread Size 12 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.