Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Chloride, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
2.5kg
Packaging:
Poly Bottle
10125-13-0
Cl2CuH4O2
170.476
MPTQRFCYZCXJFQ-UHFFFAOYSA-L
coppertrace,copper ii chloride dihydrate,copper chloride dihydrate,copper ii chloride dihydrate,caswell no. 235a,copper ii chloride dihydrate,copper 2+ chloride dihydrate,cupric chloride usp,copper chloride cucl2 , dihydrate,epa pesticide chemical code 023701
61482
CHEBI:86318
dichlorocopper;dihydrate
O.O.Cl[Cu]Cl
10125-13-0
Cl2CuH4O2
170.476
MPTQRFCYZCXJFQ-UHFFFAOYSA-L
coppertrace,copper ii chloride dihydrate,copper chloride dihydrate,copper ii chloride dihydrate,caswell no. 235a,copper ii chloride dihydrate,copper 2+ chloride dihydrate,cupric chloride usp,copper chloride cucl2 , dihydrate,epa pesticide chemical code 023701
61482
CHEBI:86318
dichlorocopper;dihydrate
O.O.Cl[Cu]Cl

High quality chemicals for laboratory and specialized industrial use

Formula Weight 170.48
Physical Form Crystals
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 2.54kg
Quantity 500g
Chemical Name or Material Cupric Chloride, Dihydrate