Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Sulfate, Colorimetric Solution (CS), 1 mL = 62.4 mg CuSO4.5H2O, 61.9-62.9 mg/mL, Spectrum™ Chemical
SDP

Catalog No. 18601915 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-915 1 L Amber Glass Bottle
18-601-914 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-601-915 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C2391LT
Add to Cart
Add to Cart

C-239, 7758-98-7

Spectrum™ Chemical Cupric Sulfate, Colorimetric Solution (CS), 1 ml = 62.4 mg CuSO4.5H2O , also known as Copper(II) Sulfate, is an aqueous solution that contains 62.4 mg CuSO4.5H2O per ml.

Chemical Identifiers

CAS 7758-99-8 , 7732-18-5
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Color Blue
CAS Max % 6.5%, 92.5%
UN Number UN1789
Concentration 61.9-62.9 mg/mL
Grade Chemical Solution
Packaging Amber Glass Bottle
Quantity 1 L
Chemical Name or Material Cupric Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.