Learn More
Selleck Chemical LLC Antazoline Phosphate
SDP
Supplier:  Selleck Chemical LLC E016425MG
Encompass_Preferred
Antazoline Phosphate is the phosphate salt of antazoline Antazoline is a first-generation antihistaminic agent and acts as a histamine H1 receptor antagonist
Catalog No. 50-313-7578
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.