Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cupric Sulfate Pentahydrate, Purified (Copper (II) Sulfate Pentahydrate), Reagents

Catalog No. 01337304
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-304 2.5 kg
01-337-303 500 g
01-337-305 12 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-304 Supplier Reagents Holdings Llc Supplier No. C21905002.5A5
Add to Cart
Add to Cart

Cupric Sulfate Pentahydrate, Purified (Copper (II) Sulfate Pentahydrate), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7758-99-8
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym Copper(II) Sulfate Pentahydrate, Blue Vitriol
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Boiling Point >150°C
Melting Point 110°C
Color Blue
pH 4
Physical Form Solid
Quantity 2.5 kg
Grade Pure
Packaging Poly Bottle
Specific Gravity 2.286
Chemical Name or Material Cupric Sulfate Pentahydrate, Purified (Copper (II) Sulfate Pentahydrate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.