Learn More
Brady™ Curb-Side Pickup Here (COVID-19) Aluminum Sign
Catalog No. 19129707 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
14 x 10 in.
Catalog No. Dimensions (L x W)
19-129-707 14 x 10 in.
1 options
Catalog No. 19-129-707 Supplier Brady™ Supplier No. 170591
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Keep your processes running smoothly and customers informed to keep them safe by identifying curbside pickup locations.
  • This durable non-reflective aluminum material (B-555) provides lasting identification with durable inks and clear-coat protection for added fade resistance
  • Designed for both indoor and outdoor use
  • Withstands wind, rain, sunlight and high temperatures
Specifications
Material Aluminum
Mounting Type Corner Holes
Languages English
Dimensions (L x W) 14 x 10 in.
Displayed Text CURBSIDE PICKUP HERE
Color Blue on White