Learn More
Selleck Chemical LLC Curcumin 50mg 458-37-7 Diferuloylmethane
SDP
Supplier:  Selleck Chemical LLC S184850MG
Encompass_Preferred
Curcumin (Diferuloylmethane) is the principal curcuminoid of the popular Indian spice turmeric, which is a member of the ginger family (Zingiberaceae). It is an inhibitor of p300 histone acetylatransferase(IC50~25 ?M) and Histone deacetylase (HDAC); activates Nrf2 pathway and supresses the activation of transcription factor NF-?B. Curcumin induces mitophagy, autophagy, apoptosis, and cell cycle arrest with antitumor activity. Curcumin reduces renal damage associated with rhabdomyolysis by decreasing ferroptosis-mediated cell death. Curcumin exhibits anti-infective properties against various human pathogens like the influenza virus, hepatitis C virus, HIV and so on. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0651193
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.