Learn More
Abcam Curcumin, Lipooxygenase Inhibitor, Solid, 250 mg
Description
- Molecular Weight: 368.4 Da
- Biological Activity: Potent antioxidant, anti-inflammatory, analgesic, and anticancer agent; acts as a lipooxygenase and cyclooxygenase inhibitor (IC50 values are 8 and 52 μM, respectively)
- Properties: High purity flavonoid derivative; shows diverse actions including free radical scavenging, cardio and neuroprotective effects
- Permeability: Blood brain barrier permeable
- Activity: Active in vitro and in vivo
- Form: Solid
- Constituents: 100% Curcumin
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | Lipooxygenase, Cyclooxygenase |
| Content And Storage | Store at -20°C. |
| Format | Vial |
| Product Type | Antioxidant |
| Form | Solid |
| Quantity | 250 mg |
| For Use With (Application) | Lipooxygenase inhibition, Cyclooxygenase inhibition, Antioxidant research, Anti-inflammatory research, Analgesic research, Anticancer research, Cardio-protection, Neuroprotection |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.