Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Curcumin, Lipooxygenase Inhibitor, Solid, 50 mg
Description
- Molecular Weight: 368.4 Da
- Biological Activity: Potent antioxidant, anti-inflammatory, analgesic and anticancer agent. Lipooxygenase and cyclooxygenase inhibitor (IC50 values are 8 and 52 μM, respectively).
- Properties: Shows diverse range of actions including free radical scavenging activity, cardio and neuroprotective effects. Blood brain barrier permeable. Active in vitro and in vivo.
- Composition: 100% Curcumin
- Form: Solid
- CAS Number: 458-37-7
Specifications
Specifications
| Buffer | 100% Curcumin |
| Shipping Condition | Ambient |
| Target Molecule | Lipooxygenase, Cyclooxygenase |
| Product Type | Antioxidant |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Formulation | 100% Curcumin |
| Quantity | 50 mg |
| For Use With (Application) | Antioxidant, Anti-inflammatory, Analgesic, Anticancer, Lipooxygenase-Cyclooxygenase inhibitor |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?