Learn More
Med Vet International Foam Dressing Kerrafoam Gentle Border 5 X 5 inch With Border Film Backing Silicone Adhesive Square Sterile, Cwl1013, 100/Case
Supplier: Med Vet International 1189361CS
Soft silicone layer ensures gentle adhesion, excellent absorption rate and nontraumatic removal Smooth outer film layer helps to reduce shear and friction, create a water-resistant bacterial barrier, and maintain an ideal MVTR (moisture vapor transmission rate) Absorbent core and foam pad combine to lock in exudate, bacteria and MMPs – even under compression Horizontal and vertical wicking layer efficiently increases absorption capacity distributing exudate throughout the dressing
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.