Learn More
United Scientific Supplies Current Balance
SDPSCI_ED
Catalog No. S27791 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Includes:
Adjustable Counterweight
Catalog No. Includes
S27791 Adjustable Counterweight
1 options
Catalog No. S27791 Supplier United Scientific Supplies Supplier No. CBA001
Add to Cart
Add to Cart
The classic method for demonstrating and measuring the small force experienced by a conductor carrying an electric current in a magnetic field
  • Pair with solenoid and external power supply to demonstrate and measure the force on a conductor carrying an electric current in a magnetic field
  • Balance features an adjustable counterweight
  • Can hold additional masses
Specifications
Product Type Current Balance
Includes Adjustable Counterweight