Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Current Technologies Dispensers for Bio-Screen™ or Fisherbrand™ BloodBloc™ Wipes

Catalog No. 1441251A
Change view
Click to view available options
Dimensions (L x W x H):
3.5 x 9.25 x 3.5 in.
4 x 10.25 x 4 in.
Color:
Clear
White
Material:
Acrylic
Cardboard
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H) Color Material
14-412-51A 4 x 10.25 x 4 in. White Cardboard
14-412-51 3.5 x 9.25 x 3.5 in. Clear Acrylic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-412-51A Supplier Current Technologies Supplier No. BH10000
This item is not returnable. View return policy

Reduce clutter on workbench, counters, trays, carts with either clear acrylic or white cardboard dispensers; sized specifically to store BloodBloc or Bioscreen wipes

For 3 × 3 in. BloodBloc (06-670-1, -36, -49), and Bio-Screen (06-670, 14-412-52C, -52H) biohazard wipes

  • Measures 3W x 3L x 3 in.H
  • For 4 × 4 in. BloodBloc (06-670-35, -50, -5) and Bio-Screen (14-412-52E, -52D, -52G) biohazard wipes
    • Measures 4W × 4L × 5.5 in.H
    • May also be used with full-size HYPE-WIPE™ bleach towelettes, 14-412-56

    Specifications

    Product Type Cardboard Dispenser
    Color White
    Dimensions (L x W x H) 4 x 10.25 x 4 in.
    Length (English) 4 in.
    Length (Metric) 10.16 cm
    Width (English) 10.25 in.
    Width (Metric) 16.5 cm
    Height (English) 4 in.
    Height (Metric) 10.16 cm
    Material Cardboard
    For Use With (Application) Reduce clutter on workbench, counters, trays, carts
    For Use With (Equipment) For 4 x 4 in. Bio-Screen or Fisherbrand BloodBloc wipes
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.