Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Curved Forceps
SCI_ED

Catalog No. S24468 Shop All Eisco Products
Change view
Click to view available options
Length (Metric):
110 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
S24468 110 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S24468 Supplier Eisco™ Supplier No. BI0172
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

With fine-pointed curved ends.

  • Made from stainless-steel

Specifications

Product Type Curved Forceps
Autoclavable Autoclavable
Color Silver
Material Stainless Steel
Length (English) 4.3 in.
Length (Metric) 110 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.