Learn More
International Crystal CURVETTE CELL WASHER
Supplier:  International Crystal 00234061
Encompass_Preferred
cuvette Cell Washer, PE Head, PP 500mL Flask, Stopper, 1/4 in. Wide aspirator Tubing, Vacuum-operated Spray Device, for Washing, Rinsing and Drying the Interior of a UV-Vis cuvette, Cleans Rectangular Cells w/ Lightpaths of 5mm, 10mm, 20mm
Catalog No. NC9279172
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.