Description
- Comfortable - 7/16 in. closed-cell nitrile foam provides excellent anti-fatigue properties
- Clean - Anti-microbial treatment provides lifetime protection from odors and degradation
- Safe - Beveled edges provide a safe transition from floor to mat; certified high-traction by the National Floor Safety Institute (NFSI)
- Versatile - Grease/oil proof, chemical resistant, welding safe
- Easy to Clean - Flexible construction makes this mat easy to clean
- Can be autoclave sterilized
- Available with or without holes for use in wet or dry areas
Specifications
Specifications
| Style | With holes |
| Color | Black |
| Certifications/Compliance | NFSI Certified. Made in USA |
| Dimensions (L x W) | 3.2 x 12.3 ft. |
| Thickness (English) | 7/16 in. |