Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ Custom TaqMan™ Gene Expression Assays

Catalog No. A80733
Encompass_Preferred
Change view
Click to view available options
Quantity:
L (2900 Reactions), Made to Order
M (750 reactions), Made to Order
S (360 reactions), Made to Order
Label or Dye:
ABY
Cy5
FAM
JUN
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Label or Dye
A80733 L (2900 Reactions), Made to Order Cy5
43-313-48 S (360 reactions), Made to Order FAM
43-320-78 M (750 reactions), Made to Order FAM
43-320-79 L (2900 Reactions), Made to Order FAM
A80713 S (360 reactions), Made to Order FAM
A80714 M (750 reactions), Made to Order FAM
A80715 L (2900 Reactions), Made to Order FAM
A80725 S (360 reactions), Made to Order ABY
A80726 M (750 reactions), Made to Order ABY
A80727 L (2900 Reactions), Made to Order ABY
A80728 S (360 reactions), Made to Order JUN
A80729 M (750 reactions), Made to Order JUN
A80730 L (2900 Reactions), Made to Order JUN
A80731 S (360 reactions), Made to Order Cy5
A80732 M (750 reactions), Made to Order Cy5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. A80733 Supplier Applied Biosystems™ Supplier No. A80733
  • $1,810.00
    $1,448.00 / Each of 1

    Save $362.00

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Applied Biosystems Custom TaqMan Gene Expression assays are the custom version of our predesigned TaqMan assays for quantitative real-time PCR.

Applied Biosystems Custom TaqMan Gene Expression assays are the custom version of our predesigned TaqMan assays for quantitative real-time PCR. The Custom TaqMan Gene Expression assays consist of a pair of unlabeled PCR primers and a TaqMan probe with a dye label (FAM, VIC, ABY, JUN, and Cyanine 5) on the 5' end and a minor groove binder (MGB) and nonfluorescent quencher (NFQ) on the 3' end.

Features include:

  • Easy to use—just add cDNA and master mix and run the qPCR—no melt curves required
  • Specific—TaqMan assays use proprietary MGB-containing probes that can be up to 15 bases shorter than non-MGB probes, improving the specificity of the assay
  • Sensitive—TaqMan assays are ideal for measuring low levels of expression or low-abundance targets
  • Accurate—identify small fold-changes with high accuracy of quantitation
  • Gold-standard TaqMan qPCR chemistry—TaqMan assays draw on Thermo Fisher Scientific's bioinformatics assay design pipeline to help ensure high specificity and minimal cross-reactivity, even for gene variants with high sequence homology
  • Sequences provided—with every Custom TaqMan Gene Expression Assay you design and order
  • Multiplexing ready—TaqMan assays are now available with 5 different dye labels to support simultaneous detection of up to 5 targets in a single qPCR reaction. FAM, ABY, JUN, and Cyanine 5 assays are available with balanced primer/probe concentrations (Primer-Limited, 350 nM) to support consistent multiplex performance. VIC Primer-Limited assays (150 nM) are available with SKU 4448487 (S), 4448488 (M), 4448492 (L).

Approximate ship time

  • Made to order: 4–6 days in North America, 4–8 days in Europe (FAM, VIC); 4-10 days in North America, 6-14 days in Europe (ABY, JUN, Cyanine 5)

Pre-formulated assay

  • 2 unlabeled PCR primers (Standard: 900 nM each final 1X concentration; Primer-Limited: 350 nM each final 1X concentration )
  • 1 dye-labeled (FAM, VIC, ABY, JUN or Cyanine 5) TaqMan MGB probe (250 nM final 1X concentration)

TaqMan Gene Expression assays are the gold standard in real-time PCR gene expression studies, built on more than 20 years of experience. Each assay includes target primers and a sequence-specific probe optimized for the best functional performance. No additional design, optimization, or melt curve analysis is needed. Available in a wide variety of formats and species, new assay designs are constantly added to help meet your research needs. TaqMan assays have been cited in over 40,000 publications and are backed by more than 350 patents.

Recommended master mix: TaqMan Fast Advanced Master Mix (Cat. No. 4444556) or TaqPath DuraPlex 1-Step RT-qPCR Master Mix (Cat. No. A58666)

See also: Custom Plus TaqMan RNA Assays, designed using the full bioinformatic power of the TaqMan Assay design pipeline.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 60X (Primer Limited)
Content And Storage •1 tube containing 20X (XS, S, and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers).
•Store at -15°C to -25°C.
Detection Method Primer-probe
Form Liquid
Format Tube
PCR Method qPCR
Reaction Speed Fast
For Use With (Equipment) 7500 System, 7500 Fast System, 7900HT System, StepOne, StepOnePlus, ViiA 7 System, QuantStudio 3, QuantStudio 5, QuantStudio 6 Flex, QuantStudio 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio 12k Flex, QuantStudio Absolute Q Digital PCR System
Internal Probe Modification Cyanine 5 (5'), MGB (Minor Groove Binder) (3')
Label or Dye Cy5
Product Line TaqMan
Product Type Gene Expression Assay
Quantity L (2900 Reactions), Made to Order
Shipping Condition Room Temperature
For Use With (Application) Gene Expression
No. of Reactions 2900 Reactions, Made to Order
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.