Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Cut Pro™ 92773 White HyperMax™ Gloves, White Polyurethane Coating

Catalog No. 19812770 Shop All MCR Safety Products
Change view
Click to view available options
Size:
2X-Large
Large
Medium
Small
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-812-770 Medium
19-812-769 Small
19-812-771 Large
19-812-772 X-Large
19-812-773 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-812-770 Supplier MCR Safety Supplier No. 92773M
Add to Cart
Add to Cart
This item is not returnable. View return policy

White Hypermax™ shell provides cut, puncture and abrasion resistance. White polyurethane palm and fingertips provides flexibility and secure grip.

  • 15-gauge
  • White Hypermax™ engineered yarn shell
  • Dexterity and flexibility
  • White PU palm and fingertip coating
  • Abrasion, cut and puncture resistant
  • Comfortable and easy to wear

Specifications

ANSI Abrasion Rating ANSI 4
Coating Material Polyurethane
Color White
Cut Resistance Rating ANSI A4
Hand Specific
Material HPPE
Product Type Cut-resistant Gloves
Size Medium
For Use With (Application) Assembly production where extreme sense of touch is needed
Glove Cuff Style Knit
Model 92773M
Puncture Rating ANSI/ISEA 105-16 Level 3
Series Cut Pro™ 92773
Yarn Gauge 15
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.