Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Cut Pro™ 9828PU ARX™ Aramid Gloves, Polyurethane Coating

Catalog No. 19812767 Shop All MCR Safety Products
Change view
Click to view available options
Size:
2X-Large
Large
Medium
Small
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-812-767 X-Large
19-812-764 Small
19-812-765 Medium
19-812-766 Large
19-812-768 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-812-767 Supplier MCR Safety Supplier No. 9828PUXL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Seamless 18-gauge green ARX™ aramid shell cut-resistant gloves feature a durable gray polyurethane coated palm and fingertips for added durability, abrasion resistance and grip.

  • 18-gauge
  • Green ARX™ shell
  • Dexterity and flexibility
  • Gray PU palm and finger coating
  • Good abrasion resistance and grip

Specifications

ANSI Abrasion Rating ANSI 3
Coating Material Polyurethane
Color Multicolored
Cut Resistance Rating ANSI A5
Hand Specific
Material HPPE
Product Type Cut-resistant Gloves
Size X-Large
For Use With (Application) Connecting metal components, fabricating duct work, sheet metal fab
Glove Cuff Style Knit
Model 9828PUXL
Puncture Rating ANSI/ISEA 105-16 Level 2
Series Cut Pro™ 9828PU
Yarn Gauge 18
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.