Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Cutibacterium acnes Derived from ATCC™ 6919™
SDP

Catalog No. 01673282 Shop All Microbiologics Products
Change view
Click to view available options
Species:
Cutibacterium acnes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species
01-673-282 Cutibacterium acnes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-673-282 Supplier Microbiologics™ Supplier No. 0170A
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Quantitative microorganism preparation kit for Cutibacterium acnes.

  • Delivers less than 100 CFU/0.1 mL of hydrated suspension
  • No dilutions required
  • Kit contains 5 pellets of a single enumerated strain and 5 vials of hydrating fluid (1.2 mL/Vial)

Specifications

ATCC® ATCC 6919
Biosafety Level 1
Format Kit
Manufacturing Origin United States
Product Type Media Test
For Use With (Application) Growth Promotion Testing, Media Testing
Quantity 50 Tests/Kit
Species Cutibacterium acnes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.