Learn More
Fireflysci Type 1 Macro Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath: 2mm)
Supplier: Fireflysci 1G2
Type 1 Macro Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath: 2mm)• Cuvette has an open top with a PTFE lid which rests on the top• Material is Optical Glass (transmits over 80% between 340-2,500nm)• Two windows optically polished• Lightpath is 2mm• Outside dimensions 4.5x12.5x45mm• Inside Dimensions- 2x10x45mm• Capacity- 0.7mL
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.