Learn More
Thomas Scientific Disposable Cuvettes, Acrylic (UV grade, useable to 275nm), Volume: 2.5ml, 2 Clear Sides, Qty: 500
SDP
Supplier:  Thomas Scientific 1158A07
Encompass
10mm path length UV grade acrylic Volume: 2.5ml 2 Clear Sides Qty: 500
Catalog No. 50-146-1597
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.