Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CUX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106971
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106971 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106971 Supplier Invitrogen™ Supplier No. PA5106971
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

CUX2 may be a transcription factor involved in neural specification. It binds to DNA in a sequence-specific manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CUX2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene CUX2
Gene Accession No. P70298
Gene Alias 1700051K22Rik; CDP2; cut like homeobox 2; Cutl2; cut-like 2; cut-like homeobox 2; CUX2; Cux-2; ENSMUSG00000072641; homeobox protein cut-like 2; Homeobox protein cux-2; KIAA0293
Gene Symbols CUX2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal region of Mouse Cux2 (aa 126-175).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13048
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.