Learn More
Abcam CX 546, AMPA Receptor Potentiator, >99% Purity, Solid, 10 mg
Description
- Molecular Weight: 247.29 Da
- Purity: >99%
- Form: Solid
- Activity: AMPA receptor potentiator; selectively binds to the agonist bound non-desensitized receptor; destabilizes desensitized receptor conformation with greater potency than CX516; enhances cognitive function in vivo
- Chemical Name: (2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)(piperidin-1-yl)methanone
- CAS Number: 215923-54-9
- Storage: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Content And Storage | Store at -20°C. |
| Target Molecule | AMPA Receptor |
| Form | Solid |
| Shipping Condition | Ambient |
| Quantity | 10 mg |
| For Use With (Application) | AMPA receptor potentiation, Cognitive function research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.