Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam CX 546, AMPA Receptor Potentiator, Solid, 50 mg

Catalog No. 502704950
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4950 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4950 Supplier Abcam Supplier No. AB14612350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AMPA receptor potentiator with >99% purity that enhances cognitive function in vivo.

  • Molecular weight: 247.29 Da
  • Purity: >99%
  • Description: AMPA receptor potentiator that selectively binds to the agonist bound non-desensitized receptor.
  • Function: Destabilizes desensitized receptor conformation with greater potency than CX516 and enhances cognitive function in vivo.
  • Form: Solid
  • Storage: Store at -20°C.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Form Solid
Shipping Condition Ambient
Target Molecule AMPA Receptor
Content And Storage Store at -20°C.
Formulation 100% CX 546
Quantity 50 mg
For Use With (Application) AMPA receptor potentiation, Cognitive function research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.