Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CX3CL1 (Fractalkine) Polyclonal Antibody, eBioscience™
GREENER_CHOICE

Catalog No. 5013714
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-137-14 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-137-14 Supplier Invitrogen™ Supplier No. 14798681
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Description: The rabbit polyclonal antibody reacts with recombinant and natural mouse, rat, and human fractalkine (CX3CL1); the antibody was generated using E. coli-expressed chemotactic domain of human CX3CL1 as an immunogen. CX3CL1, also known as neurotactin or fractalkine, is a membrane-bound CX3C chemokine. The mature protein is part of a 397-amino acid precursor and consists of a chemokine domain of 76 amino acids, a mucin stalk of 241 amino acids, a putative transmembrane domain of 18 amino acids, and an intracellular tail of 37 amino acids. Within the chemokine domain, the first two cysteine residues are separated by 3 amino acids (CX3C). CX3CL1 message is found in high abundance in the brain, kidney, lung and heart tissues. CX3CL1 is chemotactic for monocytes and other leukocytes including NK cells and may play a role in brain inflammation. Applications Reported: This polyclonal has been reported for use in immunoblotting (WB), immunoprecipitation, and immunohistochemistry. Cross-reactivity to CX3CL1 of other species has not been determined. Applications Tested: To achieve best results, titrate antibody by dilution for each application as follows: Western Blot 1:1000-1:5000 Immunoprecipitation 1:300-1:800 Immunohistochemistry 1:100-1:500. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

Chemokines are a family of proteins associated with the trafficking of leukocytes in immune surveillance and inflammatory cell recruitment. They are classified based on the positions of key cysteine residues. CX3CL1 is a CX3C chemokine known to induce adhesion and migration of leukocytes mediated by a membrane-bound and soluble form respectively. Recent experiments have shown that CX3CL1 can suppress the production of nitrous oxide, interleukin-6, and TNF-a in activated microglia and neuronal cells, suggesting that it may act as an intrinsic inhibitor against neurotoxicity by activated microglia. Its receptor, CX3CR1, also functions as a co-receptor for HIV-1 and HIV-2 envelope fusion and virus infection, which can be inhibited by CX3CL1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CX3CL1 (Fractalkine)
Applications Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Cx3cl1
Gene Accession No. O35188, O55145, P78423
Gene Alias A-152E5.2; AB030188; ABCD-3; Acc1; AI848747; C3Xkine; chemokine (C-X3-C motif) ligand 1; CX3C; CX3C membrane-anchored chemokine; C-X3-C motif chemokine 1; C-X3-C motif chemokine ligand 1; Cx3cl1; CXC3; CXC3C; D8Bwg0439e; Fkn; Fractalkine; neurotactin; NTN; NTT; Processed fractalkine; SCYD1; small inducible cytokine subfamily D (Cys-X3-Cys), member 1 (fractalkine, neurotactin); small inducible cytokine subfamily D (Cys-X3-Cys), member-1; small inducible cytokine subfamily D, 1; small-inducible cytokine D1
Gene Symbols Cx3cl1
Host Species Rabbit
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20312, 6376, 89808
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.