Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CX3CR1 Polyclonal Antibody, Invitrogen™

Catalog No. 01674784
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-784 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-784 Supplier Thermo Scientific Supplier No. PA112541

Rabbit Polyclonal Antibody

A suggested positive control for this product is human spleen lysate.

CX3CR1 is one of the chemokine receptors that are required as coreceptors for HIV infection. The genes encoding human, mouse, and rat CX3CR1 were cloned and designated V28 and CMKBRL1, CX3CR1, and RBS11, respectively. The encoded seven transmembrane protein was recently identified as the receptor for a novel transmembrane molecule, fractalkine, and renamed CX3CR1. Recently, CX3CR1 was found to serve as a coreceptor for HIV-1 and HIV-2 envelope fusion and virus infection, which can be inhibited by fractokine. CX3CR1 mediates leukocyte migration an d adhesion. CX3CR1 is expressed in a variety of human tissues and cell lines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CX3CR1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CX3CR1
Gene Accession No. P35411, P49238, Q9Z0D9
Gene Alias Beta chemokine receptor-like 1; CCRL1; chemokine (C-C) receptor-like 1; chemokine (C-X3-C motif) receptor 1; chemokine (C-X3-C) receptor 1; CMKBRL1; CMK-BRL1; CMK-BRL-1; CMKDR1; Cx3c; CX3C chemokine receptor 1; C-X3-C CKR-1; C-X3-C motif chemokine receptor 1; Cx3cr1; Fractalkine receptor; G protein-coupled receptor 13; GPR13; G-protein coupled receptor 13; GPRV28; RBS11; Scyd1; V28
Gene Symbols CX3CR1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues 2-21 of human CX3CR1 receptor.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13051, 1524, 171056
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.