Learn More
Medchemexpress LLC CXADR Mouse Biotin 20ug
SDP
Supplier:  Medchemexpress LLC HYP7236420UG
Encompass_Preferred
CXADR protein is a key component of the epithelial apical junction complex which maintains the integrity of tight junctions and enables leukocyte migration through adhesive interactions with JAML This interaction activates - T cells and promotes tissue repair through cell signaling involving PI3 kinase and MAP kinase CXADR Protein Mouse (Biotinylated HEK293 Avi-His) is the recombinant mouse-derived CXADR protein expressed by HEK293 with C-Avi C-6 His labeled tag
Catalog No. 50-001-81256