Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CXCL10/IP-10/CRG-2 Antibody Pair [HRP]
SDP

Catalog No. NB451141 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB451141 5 Plates
NB451142 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB451141 Supplier Novus Biologicals™ Supplier No. NBP3269155PLATES
Add to Cart
Add to Cart

Antibody Pair

Specifications

Assay Range 5.47-350 pg/mL (example only; lot dependent)
Assay Sensitivity 5.47 pg/mL (example only; lot dependent)
Conjugate HRP
Product Type Sandwich ELISA
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 3627
Gene Symbol CXCL10
Quantity 5 Plates
Research Discipline Biologically Active Proteins, Cancer, Cell Cycle and Replication, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Signal Transduction
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym C7, chemokine (C-X-C motif) ligand 10, crg-2, gIP-10, IFI10, INP10, IP-10, mob-1, SCYB10
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.