Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CXCL10/IP-10/CRG-2 Antibody Pair [HRP]
SDP

Catalog No. NB451142 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB451142 15 Plates
NB451141 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB451142 Supplier Novus Biologicals™ Supplier No. NBP32691515PLATES

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Antibody Pair

specifically detects in Human samples. It is validated for ELISA∣Sandwich ELISA.

Specifications

Assay Range 5.47-350 pg/mL (example only; lot dependent)
Assay Sensitivity 5.47 pg/mL (example only; lot dependent)
Conjugate HRP
Product Type Sandwich ELISA
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 3627
Gene Symbol CXCL10
Quantity 15 Plates
Research Discipline Biologically Active Proteins, Cancer, Cell Cycle and Replication, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Signal Transduction
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym C7, chemokine (C-X-C motif) ligand 10, crg-2, gIP-10, IFI10, INP10, IP-10, mob-1, SCYB10
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.