Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CXCL13 Monoclonal Antibody (DS8CX13), APC, eBioscience™, Invitrogen™

Catalog No. 17798180
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-798-180 25 μg
17-798-182 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-798-180 Supplier Invitrogen™ Supplier No. 17798180
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: This DS8CX13 monoclonal antibody recognizes mouse CXCL13 also known as BCA-1 and Angie. Applications Reported: This DS8CX13 antibody has been reported for use in flow cytometric analysis. Applications Tested: This DS8CX13 antibody has been tested by flow cytometric analysis of resident peritoneal macrophages. This may be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser

CXCL13, also known as BCA-1 and Angie, is a small chemokine strongly expressed in the secondary lymphoid organs. It preferentially promotes the migration of B lymphocytes (both B-1 and B-2) to the follicles of the spleen, lymph nodes, and Peyer's patches. It is produced by macrophages, and in mouse the highest constitutive producers include large peritoneal macrophages, Kuppfer cells and lung macrophages (both CD11c+ and CD11c-). The cellular ligand for CXCL13 is CXCR5 also known as BLR-1 (Burkitt's Lymphoma Receptor 1). CXCR5 is expressed on B cells, Tfh cells and, to a lower degree, some memory T cell populations. CXCL13 plays an important role in the development of chronic lymphocytic leukemia (CCL). CXCL13 is also a plasma biomarker for germinal center activity and generation of broadly neutralizing antibodies against HIV.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCL13
Applications Flow Cytometry
Classification Monoclonal
Clone DS8CX13
Concentration 0.2 mg/mL
Conjugate APC
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Cxcl13
Gene Accession No. O55038
Gene Alias 4631412M08Rik; Angie; ANGIE2; B cell-attracting chemokine 1; b lymphocyte chemoattractant; BCA1; BCA-1; B-cell chemoattractant; B-cell-attracting chemokine 1; B-cell-homing chemokine (ligand for Burkitt's lymphoma receptor-1); BLC; BLR1L; B-lymphocyte chemoattractant; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chem; chemokine (C-X-C motif) ligand 13; chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant); CXC; CXC chemokine BLC; CXC motif chemokine; CXC motif chemokine 13; C-X-C motif chemokine 13; C-X-C motif chemokine ligand 13; CXCL; Cxcl13; SCYB13; small inducible cytokine B subfamily (Cys-X-Cys motif), member 13 (B-cell chemoattractant); small inducible cytokine subfamily B (Cys-X-Cys), member 13; small-inducible cytokine B13
Gene Symbols Cxcl13
Host Species Rat
Immunogen Full length CXCL13.
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55985
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.