Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL17 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501731990 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-1990 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-1990 Supplier Proteintech Group Inc Supplier No. 181081AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Chemokine that acts as chemoattractant for monocytes, macrophages and dendritic cells. Plays a role in angiogenesis and possibly in the development of tumors. Acts as an anti-inflammatory in the stomach. May play a role in the innate defense against infections. Activates the C-X-C chemokine receptor GPR35 to induce a rapid and transient rise in the level of intracellular calcium ions. 4-Cys CXCL17: seems exhibit much higher chemoattractant potency on monocytes and macrophages than 6-Cys CXCL17. .

Specifications

Antigen CXCL17
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CXCL17
Gene Accession No. Q6UXB2
Gene Alias C X C motif chemokine 17, CXCL17, Dcip1, DMC, UNQ473, VCC 1, VCC1, VEGF co regulated chemokine 1
Gene Symbols CXCL17
Host Species Rabbit
Immunogen CXCL17 Fusion Protein Ag12516
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284340
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.