Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CXCL8/IL-8 Antibody Pair [HRP]
SDP

Catalog No. NB009915 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB009915 15 Plates
NB009916 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB009915 Supplier Novus Biologicals™ Supplier No. NBP27943815PLATES
Add to Cart
Add to Cart

Mouse Monoclonal Antibody Pair

specifically detects in Human samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Assay Range 3.91 pg/mL
Assay Sensitivity 3.91-250 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CXCL8/IL-8.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias 3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8SCYB8, Emoctakin, GCP-1TSG-1, interleukin 8, K60, LECT, MDNCFb-ENAP, member 8, MONAPGCP1, NAP-1NAP1, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor
Gene ID (Entrez) 3576
Gene Symbol CXCL8
Host Species Mouse
Kit Contents
  • Standard: Each vial contains 47 ng of recombinant Human CXCL8/IL-8.
  • Capture Antibody: 1 mg/mL of mouse anti-Human CXCL8/IL-8 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of mouse anti-Human CXCL8/IL-8 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Angiogenesis, Biologically Active Proteins, Cancer, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Signal Transduction
    Storage Requirements Storage is content dependent.
    Target CXCL8/IL-8
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml standard dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 250 pg/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.