Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CXCL8/IL-8 ELISA Development Kit
SDP

Catalog No. NB397884 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit (for 6 Plates)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397884 1 Kit (for 6 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB397884 Supplier Novus Biologicals™ Supplier No. NBP311755
Add to Cart
Add to Cart

ELISA Development Kit

Specifications

Assay Range 8 - 800 pg/mL
Product Type ELISA Development Kit
Sample Type Serum, Plasma, Cell Culture Supernatant
For Use With (Application) ELISA
Gene ID (Entrez) 3576
Gene Symbol CXCL8
Quantity 1 Kit (for 6 Plates)
Research Discipline Angiogenesis, Biologically Active Proteins, Cancer, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Signal Transduction
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym 3-10°C, AMCF-I, C-X-C motif chemokine 8, CXCL8SCYB8, Emoctakin, GCP-1TSG-1, interleukin 8, K60, LECT, MDNCFb-ENAP, member 8, MONAPGCP1, NAP-1NAP1, Neutrophil-activating protein 1, Protein 3-10°C, T cell chemotactic factor, T-cell chemotactic factor
Target Non-human Primate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.