Learn More
Description
It is also expected to react with chimpanzee, gorilla, and rabbit due to sequence homology. Predicted molecular weight: 42 kDa.
Specifications
Specifications
| Antigen | CXCR1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene | CXCR1 |
| Gene Accession No. | P25024, Q810W6 |
| Gene Alias | C Cmotif chemokine; C X C motif chemokine; C-C; CC motif chemokine; C-C-CKR-1; CCmotif chemokine; CD128; CD181; CDw128a; chemokine (C-X-C motif) receptor 1; CKR-1; CMKAR1; CXC; CXC chemokine receptor 1; C-X-C chemokine receptor type 1; CXC motif chemokine; C-X-C motif chemokine receptor 1; CXCR1; CXC-R1; CXCR-1; High affinity interleukin-8 receptor A; IL-8 receptor; IL-8 Receptor alpha; IL-8 Receptor IL-RA; IL-8 receptor type 1; IL-8R A; IL8R1; Il8ra; IL8RBA; IL-RA; interleukin 8 receptor; interleukin 8 receptor alpha; interleukin 8 receptor, alpha; interleukin-8 receptor type 1; interleukin-8 receptor type A |
| Gene Symbols | CXCR1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
