Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CXCR7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA3069 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA3069 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA3069 Supplier Invitrogen™ Supplier No. PA3069
  • $599.98
    $461.65 / Each of 1

    Save $138.33

    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA3-069 detects CXC Chemokine Receptor Type 7 from human and mouse samples. PA3-069 has been successfully used in Western blot and immunocytochemistry procedures. By Western blot, this antibody detects a 42 kDa protein corresponding to mouse CXC Chemokine Receptor Type 7. The PA3-069 antigen is the COOH-terminal amino acids of mouse CXCR7 (347-362) SRVSETEYSALEQNTK coupled to KLH.

This gene encodes a member of the G-protein coupled receptor family. Although this protein was earlier thought to be a receptor for vasoactive intestinal peptide (VIP), it is now considered to be an orphan receptor, in that its endogenous ligand has not been identified. The protein is also a coreceptor for human immunodeficiency viruses (HIV). Translocations involving this gene and HMGA2 on chromosome 12 have been observed in lipomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCR7
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 0.02% sodium azide
Gene Ackr3
Gene Accession No. O89039, P25106, P56485
Gene Alias ACKR3; Atypical chemokine receptor 3; AW541270; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine; chemokine (C-X-C motif) receptor 7; chemokine orphan receptor 1; Cmkor1; CXC; C-X-C chemokine receptor type 7; CXC motif chemokine; CXCR7; CXC-R7; CXCR-7; G protein-coupled receptor; GPR159; G-protein coupled receptor 159; G-protein coupled receptor RDC1 homolog; RDC1; RDC-1
Gene Symbols Ackr3
Host Species Rabbit
Immunogen COOH-terminal amino acids of mouse CXCR7 (347-362) SRVSETEYSALEQNTK coupled to KLH.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12778, 57007, 84348
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.