Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CXorf67 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552041
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552041 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552041 Supplier Invitrogen™ Supplier No. PA552041
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TVSSQASPSG GAALSSSTAG SSAAAATSAA IFITDEASGL PIIAAVLTER HSDRQDCRSP HEVFGCVVPE GGSQAAVGPQ KATGHADEHL AQTKSPGNSR RRKQPCRNQA APAQKPPGRR LFPEPLPPSS PGFRPSSYPC SGAST Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 27%.

Inhibits PRC2/EED-EZH2 complex function thereby causing down-regulation of H3K27me3. In the PRC2/EED-EZH2 complex, interacts and inhibits EZH2 methyltransferase activity. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXorf67
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CXorf67
Gene Accession No. Q86X51
Gene Alias chromosome X open reading frame 67; CXorf67; EZH inhibitory protein; EZHIP; uncharacterized protein CXorf67
Gene Symbols CXorf67
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CXorf67. Recombinant protein control fragment (Product #RP-92267).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340602
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.