Learn More
Selleck Chemical LLC CY-09 5mg 1073612-91-5
SDP
Supplier:  Selleck Chemical LLC S57745MG
Encompass_Preferred
CY-09 is a specific NLRP3 inflammasome inhibitor that directly targeted NLRP3 itself? It is tested against the five major cytochrome P450 enzymes 1A2, 2C9, 2C19, 2D6, and 3A4 with half maximal inhibitory concentration (IC50) values of 18.9, 8.18, >50, >50, and 26.0 ¿M, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8315
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.