Learn More
Description
CyDye™ fluors are versatile fluorophores designed for a broad range of applications. CyDye™ fluors produce high sensitivity and highly reproducible performance at a biologically suitable pH range of 3 to 10.
Cy™3B Maleimides is a more selective method of antibody labeling that reduces the possibility of the label interfering with subsequent antibody-antigen reactions.
- The use of multicolor CyDye™ flours enables multiplexing without crosstalk for high accuracy
- CyDye™ flours readily dissolve in aqueous buffers, which eliminates the need of organic solvents
- High purity and thorough QC testing generate high levels of chromophore and reactive dye content
- Narrow excitation and emission bands result in discrete signals from each fluor
Specifications
Specifications
| Formulation | 100% Cy3B (maleimide) |
| Format | Solid |
| Quantity | 1 mg |
| Product Type | Cy3B Maleimide |
| Molecular Weight (g/mol) | 796 |
| Solubility | Soluble in cold and hot water |
| Label or Dye | Cy3B |
| Color | Red |
| Emission | 570 |
| Excitation | 559 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.