Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Cy™5.5 Amidite

Catalog No. 09928192 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-928-192 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-928-192 Supplier Cytiva Supplier No. 28904250
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For CyDye™ incorporation at any position in an oligonucleotide during synthesis

CyDye™ fluors combine exceptional photostability with bright and intense signals, resulting in high sensitivity and a highly reproducible performance at a biologically suitable pH range of 3 to 10.

  • Wide selection of CyDye™ fluors for protein labeling for use in ELISA & immunoprecipitation
  • Labeling strategy based on protein characteristics
  • Medium to high-throughput fluorescence imaging
  • Ready-to-use labeled antibodies enable multiplexed assays
  • Streptavidin-conjugated CyDye™ fluors for use in Flow-cytometry and In situ hybridization
  • Linkage of Cy dyes allows labeled DNA to hybridize to other DNA
  • DNA synthesis reagent

Amersham™ CyDye™ fluors are versatile fluorophores designed for a broad range of applications. The use of multicolor CyDye™ fluors enables multiplexing without crosstalk. In addition, the CyDye™ flours readily dissolve in aqueous buffers, which eliminates the need of organic solvents. The high purity and thorough QC testing of Amersham™ CyDye™ fluors generate high levels of chromophore and reactive dye content. Narrow excitation and emission bands result in discrete signals from each fluor, which together with minimal cross-talk contributes to high accuracy.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, away from incompatible materials
Description Stable
Label or Dye Cy5.5
Product Line Cy
Product Type Cy5.5 Amidite
Quantity 100 mg
Size 100 mg
Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.