Learn More
Apexbio Technology LLC Cy3 alkyne (non-sulfonated), 25mg. Cas: MFCD: NA. used for Click chemistry to label any molecule bearing azide group.
SDP
Supplier:  Apexbio Technology LLC A813025
Encompass_Preferred
Cy3 alkyne is a water-insoluble dye ready for the use in Click Chemistry reaction. With this bright and photostable reagent, molecules bearing azide group can be attached by it. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5040
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.