Learn More
Cytiva Cy™3 Amidite
Catalog No. 09928190 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 mg
Catalog No. Quantity
09-928-190 100 mg
1 options
Catalog No. 09-928-190 Supplier Cytiva Supplier No. 28917298
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
For CyDye™ incorporation at any position in an oligonucleotide during synthesis
CyDye™ fluors combine exceptional photostability with bright and intense signals, resulting in high sensitivity and a highly reproducible performance at a biologically suitable pH range of 3 to 10.
  • Wide selection of CyDye™ fluors for protein labeling for use in ELISA & immunoprecipitation
  • Labeling strategy based on protein characteristics
  • Medium to high-throughput fluorescence imaging
  • Ready-to-use labeled antibodies enable multiplexed assays
  • Streptavidin-conjugated CyDye™ fluors for use in Flow-cytometry and In situ hybridization
  • Linkage of Cy dyes allows labeled DNA to hybridize to other DNA
  • DNA synthesis reagent
Amersham™ CyDye™ fluors are versatile fluorophores designed for a broad range of applications. The use of multicolor CyDye™ fluors enables multiplexing without crosstalk. In addition, the CyDye™ flours readily dissolve in aqueous buffers, which eliminates the need of organic solvents. The high purity and thorough QC testing of Amersham™ CyDye™ fluors generate high levels of chromophore and reactive dye content. Narrow excitation and emission bands result in discrete signals from each fluor, which together with minimal cross-talk contributes to high accuracy.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) For incorporation of Cy3 fluorescent dyes at any position in an oligonucleotide during synthesis
Product Type Cy3 Amidite
Quantity 100 mg