Learn More
Apexbio Technology LLC Cy3 hydrazide (non-sulfonated), 25mg. Cas: MFCD: NA. carbonyl-reactive dye
SDP
Supplier:  Apexbio Technology LLC A826525
Encompass_Preferred
Cy3 hydrazide is a carbonyl-reactive dye, an analog of Cy3 hydrazide. Cyanine3 dyes have been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5099
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.