Learn More
Apexbio Technology LLC Cy5 carboxylic acid, 25mg. Cas: MFCD: NA. Non-reactive fluorophore, for control experiments, and for calibration
SDP
Supplier:  Apexbio Technology LLC A813725
Encompass_Preferred
Sulfo-Cy5 carboxylic acid is a non-activated, sulfonated, hydrophilic and highly water soluble dye. Non-Sulfo-Cy5 carboxylic acid analog is available. For kinds of researches related experiments, using pre-activated sulfo-Cy5 NHS ester is preferred Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5050
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.