Learn More
Apexbio Technology LLC Cy5 NHS ester, 5mg. Cas: MFCD: MFCD28142487. Labeling of amino-groups in biomolecules.
SDP
Supplier:  Apexbio Technology LLC A81085
Encompass_Preferred
Sulfo-Cy5 NHS ester is a sulfonated, hydrophilic and highly water soluble dye. For the labeling of various amine-containing molecules in aqueous phase, no organic co-solvent is needed. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5001
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.