Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Cy7 Bis NHS Ester
GSA_VA

Catalog No. 45000483 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-000-483 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-000-483 Supplier Cytiva Supplier No. PA17002

missing translation for 'validMainframeMsg'

Suitable for general labeling of amine residues

The packs contain fluors carrying two reactive groups on each dye molecule for general labeling of protein amine groups.
  • Suitable for general labeling of amine residues.
  • Large range of bulk packs available for greatest economy.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Formulation 100% Cy7 (Bis NHS ester)
For Use With (Application) Used for high-throughput screening (HTS) applications
Quantity 50 mg
Product Type Cy7 Bis NHS Ester
Content And Storage 2°C to 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles.
Label or Dye Cy7
Color Purple
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.