Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyanide Standard, Certified, 1000ppm ±10ppm (1mL = 1mg CN-), LabChem™

Catalog No. LC135451
Change view
Click to view available options
Quantity:
125 mL
500 mL
1 L
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC135451 500 mL Poly Bottle
LC135457 125 mL Poly Bottle
LC135452 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC135451 Supplier LabChem, Inc. Supplier No. LC135451
Add to Cart
Add to Cart

Chemical Identifiers

CAS 151-50-8 , 1310-73-2
Molecular Formula CKN
Molecular Weight (g/mol) 65.116
InChI Key NNFCIKHAZHQZJG-UHFFFAOYSA-N
PubChem CID 9032
IUPAC Name potassium;cyanide
SMILES [C-]#N.[K+]

Specifications

Packaging Poly Bottle
CAS Max % 99.55, 0.25, 0.2
Identification Passes Test
Density 1g/mL
Physical Form Liquid
Concentration 1000 ppm ±10 ppm (1mL = 1mg CN-)
Linear Formula KCN
Color Colorless
Solubility Soluble in water
Quantity 500 mL
Formula Weight 65.12
Grade Certified
Chemical Name or Material Cyanide Standard, 1000ppm (1mL = 1mg CN-)
Show More Show Less
Hazard Statement GHS H Statement
Harmful to aquatic life.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.