Learn More
Medchemexpress LLC HY-135414B 5mg Medchemexpress, Cyanine5 NHS ester (iodide) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY135414B5MG
Encompass_Preferred
Medchemexpress, HY-135414B 5mg Cyanine5 NHS ester (iodide) CAS: Cyanine5 NHS ester iodide is a red emitting fluorescent dye for labeling of amino-groups in peptides, proteins, and oligonucleotides. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1389
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.