Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyanogen bromide, 97%

Catalog No. AC110781000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle in Tin Can
Glass Bottle packed in tin can
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC110781000 100 g Glass Bottle in Tin Can
AC110780250 25 g Glass Bottle in Tin Can
AC110785000 500 g Glass Bottle packed in tin can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC110781000 Supplier Thermo Scientific Chemicals Supplier No. 110781000
Add to Cart

CAS: 506-68-3 | CBrN | 105.922 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-68-3
Molecular Formula CBrN
MDL Number MFCD00011597
InChI Key ATDGTVJJHBUTRL-UHFFFAOYSA-N
Synonym cyanogen bromide, bromine cyanide, bromocyanide, cyanobromide, bromocyanogen, bromocyan, campilit, cyanogen monobromide, bromure de cyanogen, cyanogen bromide brcn
PubChem CID 10476
IUPAC Name carbononitridic bromide
SMILES C(#N)Br

Specifications

Color White
Density 1.44
Assay Percent Range 97%
Packaging Glass Bottle in Tin Can
Linear Formula BrCN
Quantity 100 g
Fieser 01,174; 02,93; 04,110; 05,169; 06,148; 08,131; 09,127
Merck Index 15, 2683
Molecular Weight (g/mol) 105.922
Formula Weight 105.93
Percent Purity ≥96.0%
Physical Form Crystals
Chemical Name or Material Cyanogen bromide, 97%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Recommended Storage Refrigerator +4°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.